Anti-CD3e Mouse Monoclonal Antibody [clone: UCHT1] (APC/iFluor® 750)

Lieferant: AAT BIOQUEST
100321G0 100321G1 100321G2
AATB100321G0EA 319 EUR
AATB100321G0 AATB100321G1 AATB100321G2
Anti-CD3e Mouse Monoclonal Antibody [clone: UCHT1] (APC/iFluor® 750)
Antikörper
The UCHT1 monoclonal antibody binds to human CD3e, a 20 kD member of the Ig superfamily commonly found on the surface of tregs, thymocytes, nkt cells and thymocytes (differentiation dependents).

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Bitte beachten For research use only.

Type: Primary
Antigen: CD3e
Clonality: Monoclonal
Clone: UCHT1
Conjugation: APC/iFluor® 750
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR