Anti-Rabbit IgG Goat Polyclonal Antibody (CF®505)

Lieferant: Biotium
20915-50UL 20915-250UL
20915-50ULEA 103 EUR
20915-50UL 20915-250UL
Anti-Rabbit IgG Goat Polyclonal Antibody (CF®505)
Antikörper
This is a goat anti-rabbit IgG (H L) F(ab’)2 fragment secondary antibody labeled superior CF® Dyes.

Type: Secondary
Antigen: Rabbit IgG
Clonality: Polyclonal
Clone:
Conjugation: CF®505
Epitope:
Host: Goat
Isotype: IgG (H+L)
Reactivity: Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR