Paroxetine hydrochloride ≥98%
Lieferant: ENZO LIFE SCIENCES
Synonyme:
(3S,4R)-3-((Benzo[d][1,3]dioxol-5-yloxy)methyl)-4-(4-fluorophenyl)piperidine hydrochloride
Highly potent and selective serotonin uptake inhibitor
Paroxetine is a highly potent and selective serotonin (5-HT) uptake inhibitor. It acts by binding to the serotonin transporter (SERT) with high affinity (Ki = 05 nM). In vitro paroxetine competitively inhibits the uptake of 5-HT by rat hypothalamic and cortical synaptosomes with only very weak effects on noradrenaline and dopamine uptake. Paroxetine is in clinical use for the treatment of depression, anxiety and other disorders.
Formel:
C₁₉H₂₁ClFNO₃ |
CAS-Nummer:
78246-49-8 |