all cis-5,8,11,14,17-Eicosapentaenoic acid ≥98% (durch TLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme: 5(Z),8(Z),11(Z),14(Z),17(Z)-Eicosapentaenoic Acid, Timnodonic acid, cis-5,8,11,14,17-Eicosapentaenoic acid, EPA
BMLFA0010100 BMLFA0011000
ENZOBMLFA0010100EA 114 EUR
ENZOBMLFA0010100 ENZOBMLFA0011000
all cis-5,8,11,14,17-Eicosapentaenoic acid ≥98% (durch TLC)
all cis-5,8,11,14,17-Eicosapentaenoic acid
Beneficial effect in cardiovascular diseases and inflammation. Inhibits PGE2 formation from arachidonic acid in 3T3 fibroblats.
  • Inhibits PGE2 formation
Colorless oil.
Soluble in 100% ethanol.
Formel: C₂₀H₃₀O₂
Molecular Weight: 302,46 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00065716
CAS-Nummer: 10417-94-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d83ac34b7997c9e32c8b830946d26f4a0d003a38&vid=b3c0b22f0766472709f2d1d8237f567869b9d7d5&prid=web&clid=SAPDAM