Seratrodast ≥98%

Lieferant: ENZO LIFE SCIENCES
Achtung

Synonyme: 7-Phenyl-7-(2,4,5-trimethyl-3,6-dioxocyclohexa-1,4-dien-1-yl)heptanoic acid

BMLRA1190010 BMLRA1190050
ENZOBMLRA1190010EA 110 EUR
ENZOBMLRA1190010 ENZOBMLRA1190050
Seratrodast ≥98%
Seratrodast

Thromboxane receptor ligand.


Seratrodast is a potent and selective thromboxane A₂ receptor (TP) antagonist with antioxidant properties.


Soluble in DMSO (12mg/ml); Ethanol (10mg/ml)

Formel: C₂₂H₂₆O₄
Molecular Weight: 354.45 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00875701
CAS-Nummer: 112665-43-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=facd2bb44ca138f65fcd31ac0386646c8958ff67&vid=9a23b76c3fd6bdf368d2a5fc9d5dee6bd396decf&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Purity ≥98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR