Tyrphostin 1 ≥99%
Lieferant: ENZO LIFE SCIENCES
Synonyme:
Tyrphostin A1, 2-(4-Methoxybenzylidene)malononitrile, ((4-Methoxyphenyl)methylene)methane-1,1-dicarbonitrile, (4-Methoxybenzylidene)malononitrile
BMLEI1900020
BMLEI1900100
ENZOBMLEI1900020EA
84.7
EUR
ENZOBMLEI1900020
ENZOBMLEI1900100
Tyrphostin 1 ≥99%
Tyrphostin 1
An inactive tyrphostin, which may be used as a negative control.
Soluble in 100% ethanol (25 mg/ml, warm) or DMSO (25 mg/ml).
Formel:
C₁₁H₈N₂O Molecular Weight: 184.198 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00019787 CAS-Nummer: 2826-26-8 |
|
Spezifikation Testergebnisse
Purity | ≥99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...