Tyrphostin A9 ≥99% (durch HPLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme: [4-Hydroxy-3,5-bis(2-methyl-2-propanyl)benzylidene]malononitrile, Malonoben
BMLEI2150050 BMLEI2150250
ENZOBMLEI2150050EA 89.1 EUR
ENZOBMLEI2150050 ENZOBMLEI2150250
Tyrphostin A9 ≥99% (durch HPLC)
Tyrphostin A9
Tyrphostin 9 is a selective inhibitor of the PDGF receptor tyrosine kinase (IC50=1.2 µM). It is also a potent (10 nM) uncoupler of oxidative phosphorylation.
  • PDGF receptor inhibitor
Yellow solid.
Soluble in DMSO (>25mg/ml) or ethanol (20mg/ml).
Formel: C₁₈H₂₂N₂O
Molecular Weight: 282.387 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00209853
CAS-Nummer: 10537-47-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3b650322c94c0ed0e241e857bb9af473ad1740fe&vid=d5153d7da389d78435ad30c86565eb697f4d917f&prid=web&clid=SAPDAM