DMOG ≥99% (durch TLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme: N-[Methoxy(oxo)acetyl]glycine methyll ester, Dimethyloxaloylglycine, Methyl 2-((2-methoxy-2-oxoethyl)amino)-2-oxoacetate, DMOG
BMLEI3470010 BMLEI3470050
ENZOBMLEI3470010EA 89.1 EUR
ENZOBMLEI3470010 ENZOBMLEI3470050
DMOG ≥99% (durch TLC)
DMOG
DMOG is a cell permeable prolyl-4-hydroxylase inhibitor which upregulates HIF activity. HIF activation stimulates angiogenesis in several different models. DMOG also inhibits FIH (Factor Inhibiting HIF), an asparaginyl hydroxylase, which enhances the HIF response. It is active in vivo and attenuates myocardial injury in a rabbit ischemia reperfusion model (20mg/kg). Is expected to act pro-angiogenic.
  • Prolyl-4-hydroxylase inhibitor
Off-white solid.
Soluble in DMSO (>25mg/ml), 100% ethanol (>25mg/ml) or dimethyl formamide; soluble also in PBS, pH7.2 (10mg/ml).
Formel: C₆H₉NO₅
Lagertemperatur: Gefrierschrank
MDL: MFCD05865098
CAS-Nummer: 89464-63-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=612af2854b6767a0f957c24790bb8de6dfaa3670&vid=cadf617133565e51b046f418036c22e2df01a397&prid=web&clid=SAPDAM