SC-51322 ≥98%
Lieferant: ENZO LIFE SCIENCES
Synonyme:
3-chloro-N'-[3-(furan-2-ylmethylsulphanyl)propanoyl]-6H-benzo[b][1,4]benzoxazepine-5-carbohydrazide
BMLRA1120005
BMLRA1120025
ENZOBMLRA1120005EA
110
EUR
ENZOBMLRA1120005
ENZOBMLRA1120025
SC-51322 ≥98%
SC-51322
Prostanoid receptor inhibitor
A potent and selective prostaglandin E2 antagonist (pA2=8.1). Displays in vivo analgesic activity in the mouse (ED50=0.9 mg/kg).
Formel:
C₂₂H₂₀ClN₃O₄S Molecular Weight: 457.929 g/mol |
MDL:
MFCD00916246 CAS-Nummer: 146032-79-3 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...