Wiskostatin ≥98%

Lieferant: ENZO LIFE SCIENCES

Synonyme: 1-(3,6-Dibromo-9H-carbazol-9-yl)-3-(dimethylamino)propan-2-ol

BMLT1260001 BMLT1260005
ENZOBMLT1260001EA 84.7 EUR
ENZOBMLT1260001 ENZOBMLT1260005
Wiskostatin ≥98%
Wiskostatin

Blocks actin filament assembly


A selective, reversible inhibitor of N-WASP (neural Wiskott-Aldrich syndrome protein) that selectively blocks actin filament assembly. Appears to bind to N-WASP, stabilize the autoinhibited conformation and prevent the activation of Arp2/3 (actin-related protein 2/3) complex.

Formel: C₁₇H₁₈Br₂N₂O
Molecular Weight: 426,152 g/mol
MDL: MFCD00218393
CAS-Nummer: 253449-04-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f635803d5165d1a0004f7bf3de3a3bef49fffbc6&vid=5f03c183cf1752f43233892aa434a920b2f6e671&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR