Cinnamyl-3,4-dihydroxy-α-cyanocinnamate ≥98%
Lieferant: ENZO LIFE SCIENCES
BMLEI2110010
BMLEI2110050
ENZOBMLEI2110010EA
110
EUR
ENZOBMLEI2110010
ENZOBMLEI2110050
Cinnamyl-3,4-dihydroxy-α-cyanocinnamate ≥98%
Cinnamyl-3,4-dihydroxy-alpha-cyanocinnamate
Lipoxygenase inhibitor
Selective 12-lipoxygenase inhibitor. IC50=0.063 µM, 3.33 µM and 1.89 µM for 12-, 15- and 5-LO respectively in isolated enzyme preparations.
Formel:
C₁₉H₁₅NO₄ Molecular Weight: 321,332 g/mol |
MDL:
MFCD00210839 CAS-Nummer: 132465-11-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...