Tyrphostin AG-126 ≥98% (durch HPLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme: 2-[(3-Hydroxy-4-nitrophenyl)methylidene]propanedinitrile, Tyrphostin AG 126, 2-(3-Hydroxy-4-nitrobenzylidene)malononitrile, AG-126, (3-Hydroxy-4-nitrobenzylidene)malononitrile
BMLEI2630005 BMLEI2630025
ENZOBMLEI2630005EA 110 EUR
ENZOBMLEI2630005 ENZOBMLEI2630025
Tyrphostin AG-126 ≥98% (durch HPLC)
Tyrphostin AG-126
AG-126 prevents lipopolysaccharide-induced lethal toxicity (septic shock) in mice. Administered at 1.5 mg/mouse, it reduced LPS-induced lethal toxicity from 95% to 10%. AG-126 greatly reduces LPS-induced tyrosine phosphorylation of p42MAPK in murine peritoneal macrophages at 50 µM. EGFR, Her-2/neu receptor or PDGF-R are not inhibited at 100 µM. Inhibits tyrosine kinase-dependent B cell receptor signaling.
  • Inhibits tyrosine kinase-dependent B cell receptor signaling
Yellow solid.
Soluble in DMSO (25mg/ml) or 100% ethanol (5mg/ml warm).
Formel: C₁₀H₅N₃O₃
Molecular Weight: 215.168 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00236448
CAS-Nummer: 118409-62-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6298223c475925f46712741e5195d64fe4a6d9ae&vid=1b6917c92d32148a7c38d78a80f41be6cb0900ee&prid=web&clid=SAPDAM