δ-methrin ≥99%

Lieferant: ENZO LIFE SCIENCES

Synonyme: (S)-alpha-Cyano-3-phenoxybenzyl (1R,3R)-3-(2,2-Dibromovinyl)-2,2-dimethylcyclopropanecarboxylate, (1R,3R)-3-(2,2-Dibromovinyl)-2,2-dimethylcyclopropanecarboxylic acid (S)-alpha-Cyano-3-phenoxybenzyl ester, Deltamethrin

BMLPR1010010 BMLPR1010050
ENZOBMLPR1010010EA 86.9 EUR
ENZOBMLPR1010010 ENZOBMLPR1010050
δ-methrin ≥99%
Deltamethrin

Calcineurin inhibitor


Synthetic type II pyrethroid. Potent calcineurin (protein phosphatase 2B) inhibitor IC50~0.03 nM. Causes an increase in neurotransmitter release at synapses and also increases intrasynaptosomal Ca2+ levels. Serious doubts have been raised about the efficacy of this inhibitor. May also have varied effects on ion channels.

Formel: C₂₂H₁₉Br₂NO₃
Molecular Weight: 505.206 g/mol
MDL: MFCD00870122
CAS-Nummer: 52918-63-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=be398de600492ac57e01fb6379dbca5cc312f802&vid=3294ee5b7eb59938cd80b7f79b918c55b92546b3&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR