Latrunculin A ≥98% (durch TLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme: (4R)-4-[(1R,4S,5E,7Z,11E,15R,17R)-17-hydroxy-4,11-dimethyl-13-oxo-14,18-dioxabicyclo[13.3.1]nonadeca-5,7,11-trien-17-yl]-1,3-thiazolidin-2-one
BMLT1190100 BMLT1190500
ENZOBMLT1190100EA 171 EUR
ENZOBMLT1190100 ENZOBMLT1190500
Latrunculin A ≥98% (durch TLC)
Latrunculin A
Ultra-pure; inhibits actin polymerization
Inhibits actin polymerization, microfilament organzation and microfilament-mediated processes. Acts via a different mechanism and 10- to 20-fold more potently than cytochalasins. Inhibits phagocytosis by macrophages. Exposure to 0.2 mg/ml for one hour produces maximal change in cell morphology.
Formel: C₂₂H₃₁NO₅S
Molecular Weight: 421.552 g/mol
CAS-Nummer: 76343-93-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6ecf1ee0ecf889b337739abc75420a9544058d88&vid=da41150ef3e1a659af05094d3a91f581919c8143&prid=web&clid=SAPDAM