PP2 ≥98% (durch TLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme: 1-(tert-Butyl)-3-(4-chlorophenyl)-1H-pyrazolo[3,4-d]pyrimidin-4-amine
BMLEI2970001 BMLEI2970005
ENZOBMLEI2970001EA 110 EUR
ENZOBMLEI2970001 ENZOBMLEI2970005
PP2 ≥98% (durch TLC)
PP2
Src kinase inhibitor
A potent and selective inhibitor of the Src family tyrosine kinases. Inhibits Lck (IC50=4 nM), Fyn (IC50=5 nM), and Hck (IC50=5 nM). It displays weaker inhibition of EGFR (IC50=480 nM) and no inhibition of ZAP-70 (IC50 100 µM) and JAK2 (IC5050 µM). PP2 is a useful tool for probing the involvement of Src family tyrosine kinases in cellular signaling.
Formel: C₁₅H₁₆ClN₅
Molecular Weight: 301,8 g/mol
MDL: MFCD01568210
CAS-Nummer: 172889-27-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c3b40f2b5783b0a0b533ee6b8f08aafef6c1a2aa&vid=5ebdd8d33b2afccf9d07a3a2232fc2fd3b01130e&prid=web&clid=SAPDAM