Dibutyryl-cAMP sodium salt ≥98% (durch HPLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme: Bucladesine sodium salt, DC-2797, Dibutyl cyclic AMP sodium salt, Dibutyryl-cAMP, N6,2'-O-Dibutyryladenosine 3',5'-Cyclic monophosphate sodium salt, DbcAMP, Sodium (4aR,6R,7R,7aR)-6-(6-butyramido-9H-purin-9-yl)-7-(butyryloxy)tetrahydro-4H-furo[3,2-d][1,3,2]dioxaphosphinin-2-olate 2-oxide, Bucladesine
BMLCN1250030 BMLCN1250100
ENZOBMLCN1250030EA 88 EUR
ENZOBMLCN1250030 ENZOBMLCN1250100
Dibutyryl-cAMP sodium salt ≥98% (durch HPLC)
Dibutyryl-cAMP sodium salt
PKA activator.
A cell-permeable cAMP analog which preferentially activates cAMP-dependent protein kinase. The products releaes butyrate due to intracellular and extracellular esterase action. Butyrate was shown to have distinct biological effects.
Soluble in water (100 mg/ml), DMSO (200 mM) or 96% ethanol (25 mM).
Formel: C₁₈H₂₃N₅O₈P·Na
Molecular Weight: 491.37 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00005843
CAS-Nummer: 16980-89-5
EINECS: 241-059-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=01751242474e209b481659378043656b9f7cae4e&vid=174f765272dc5542a1079f769a8709dfa0a865b7&prid=web&clid=SAPDAM
Bestellen
Spezifikation Testergebnisse
Purity ≥98% (HPLC)
Identity Determined by UV spectrum