L-Ergothioneine ≥98% (durch TLC), weißer Feststoff
Lieferant: ENZO LIFE SCIENCES
Synonyme: (S)-3-(2-Thioxo-2,3-dihydro-1H-imidazol-4-yl)-2-(trimethylammonio)propanoate, Ergothioneine
BMLFR1110010 BMLFR1110050
ENZOBMLFR1110010EA 165 EUR
ENZOBMLFR1110010 ENZOBMLFR1110050
L-Ergothioneine ≥98% (durch TLC), weißer Feststoff
L-Ergothioneine
Antioxidant.
  • Water soluble
Naturally occuring ubiquitous antioxidant which is found in many cells at concentrations up to 2 mM. Peroxynitrate ONOO⁻ scavenger. Protects human neuronal hybridoma cell line (N-18-RE-105) from ONOO⁻-induced oxidative damage.
Formel: C₉H₁₅N₃O₂S
Molecular Weight: 229,3 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00167474
CAS-Nummer: 497-30-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=927ad62b3350ab9f4e97037cff5ad5f71be0d393&vid=5795dc3afb92ab5292dabb56c6d614c07b5027d6&prid=web&clid=SAPDAM
Bestellen
Spezifikation Testergebnisse
Purity ≥98% (TLC)