CITCO
Lieferant: ENZO LIFE SCIENCES
|
Achtung
|
Synonyme:
6-(4-Chlorophenyl)imidazo[2,1-b]thiazole-5-carbaldehyde O-(3,4-dichlorobenzyl) oxime, 6-(4-Chlorophenyl)imidazo[2,1-b] [1,3]thiazole-5-carbaldehyde O-3,4-dichlorobenzyl) oxime
Constitutive androstane receptor ligand.
CITCO is a novel potent and selective agonist for the human orphan nuclear receptor CAR (constitutive androstane receptor). CITCO displayed an EC50 of 49 nM in a CAR FRET assay and showed 50-fold selectivity for CAR over PXR. CITCO displayed no activity against a panel of 15 other nuclear receptors at a concentration of 10 µM. In hepatocytes CITCO induced CAR nuclear translocation and induced the CAR target gene CYP2B6.
Soluble in DMF (25 mg/ml) or DMSO (10 mg/ml).
Formel:
C₁₉H₁₂Cl₃N₃OS Molecular Weight: 436,7 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00139608 CAS-Nummer: 338404-52-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...