Misoprostol ≥99% (durch TLC)
Lieferant: ENZO LIFE SCIENCES
Gefahr
Synonyme: (±)-15-Deoxy-(16RS)-16-hydroxy-16-methylprostaglandin E1 methyl ester, SC-29333
BMLPG0510010 BMLPG0510100
ENZOBMLPG0510010EA 165 EUR
ENZOBMLPG0510010 ENZOBMLPG0510100
Misoprostol ≥99% (durch TLC)
Misoprostol
Prostanoid receptor ligand.
A cytoprotective PGE₁ analog. Prevents NSAID-induced gastric ulceration. Misoprostol is a methyl ester and is therefore inactive in receptor preparations and in other situations where it does not undergo ester hydrolysis.
Soluble in ethanol (50 mg/ml).
Formel: C₂₂H₃₈O₅
Molecular Weight: 382,5 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00274550
CAS-Nummer: 59122-46-2
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6700d361d7a4c47e6900fedf6089469f3005ba99&vid=16b6752a620fe60771cd1adc0b717b53dae24927&prid=web&clid=SAPDAM
Bestellen
Spezifikation Testergebnisse
Purity ≥99% (TLC)
Identity Determined by NMR