Bimatoprost ≥97%
Lieferant: ENZO LIFE SCIENCES
|
Gefahr
|
Synonyme:
(Z)-7-((1R,2R,3R,5S)-3,5-Dihydroxy-2-((S,E)-3-hydroxy-5-phenylpent-1-en-1-yl)cyclopentyl)-N-ethylhept-5-enamide
BMLRA1210001
BMLRA1210010
ENZOBMLRA1210001EA
75.9
EUR
ENZOBMLRA1210001
ENZOBMLRA1210010
Bimatoprost ≥97%
Bimatoprost
FP receptor agonist.
Stable analog of PGF2α. Agonist at FP receptors (after in vivo hydrolysis to the free acid). It is an efficacious treatment for glaucoma. It also displays an unusual eye lash lengthening effect.
Soluble in DMSO (25 mg/ml) or ethanol (25 mg/ml).
Formel:
C₂₅H₃₇NO₄ Molecular Weight: 415,6 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD03411999 CAS-Nummer: 155206-00-1 |
Spezifikation Testergebnisse
Purity | ≥97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...