GW-441756 ≥98%

Lieferant: ENZO LIFE SCIENCES

Synonyme: GW 441756, 3-((1-Methyl-1H-indol-3-yl)methylene)-1,3-dihydro-2H-pyrrolo[3,2-b]pyridin-2-one

BMLEI3640010 BMLEI3640050
ENZOBMLEI3640010EA 211 EUR
ENZOBMLEI3640010 ENZOBMLEI3640050
GW-441756 ≥98%
GW-441756

TrkA inhibitor


Potent and selective inhibitor of NGFR kinase (TrkA, IC50=2 nM). Blocks TrkA-mediated γH2AX production and apoptosis in TrkA-overexpressing cells.

Formel: C₁₇H₁₃N₃O
Molecular Weight: 275.3 g/mol
MDL: MFCD17010281
CAS-Nummer: 504433-23-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba4206a8e51b32ac9fc098ce2a121c69715139fe&vid=f4d54967b91a04de29f891dfc7b7dcae5edc8dbe&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR