IC87114 ≥99% (durch HPLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme: 2-((6-Amino-9H-purin-9-yl)methyl)-5-methyl-3-(o-tolyl)quinazolin-4(3H)-one, IC-87114
ENZCHM1380005 ENZCHM1380010
ENZOENZCHM1380005EA 147 EUR
ENZOENZCHM1380005 ENZOENZCHM1380010
IC87114 ≥99% (durch HPLC)
IC87114
Cell permeable, selective inhibitor of PI 3-Kδ.
Formel: C₂₂H₁₉N₇O
Molecular Weight: 397,4 g/mol
MDL: MFCD09970845
CAS-Nummer: 371242-69-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df4fd13b9ab5a9572c3c80f4630fd62012a7e038&vid=f8db61decf154e1741d9121b6b87763413056104&prid=web&clid=SAPDAM