Fumonisin B2 ≥80%
Lieferant: ENZO LIFE SCIENCES
Synonyme:
1,2,3-Propanetricarboxylic acid, 1,1?-[1-(12-amino-4,11-dihydroxy-2-methyl-tridecyl)-2-(1-methylpentyl)-1,2-ethane-diyl]ester, Fumonisin B2 from Fusarium moniliforme, sphingosine N-acyltransferase inhibitor, 1,2,3-Propanetricarboxylic acid, 1,1'-(1-(2-amino-9,11-dihydroxy-2-methyltridecyl)-2-(1-methylpentyl)-1,2-ethanediyl) ester
Ceramide synthase inhibitor
Mycotoxin, structurally similar to fumonisin B1 (Prod. No. BML-SL220). Found as contaminant mostly in corn silage; was detected recently for the first time in the industrially important Aspergillus niger. Induces cancer. Inducer of apoptosis. Inhibits sphinganine N-acyltransferase (ceramide synthase) at IC50 = 0.1µM. Inhibits de novo sphingolipid biosynthesis.
Formel:
C₃₄H₅₉NO₁₄ Molecular Weight: 705,839 g/mol |
MDL:
MFCD00133350 CAS-Nummer: 116355-84-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...