PNU-74654 ≥98% (durch HPLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme: N'-((5-Methylfuran-2-yl)methylene)-2-phenoxybenzohydrazide
BMLWN1010005 BMLWN1010025
ENZOBMLWN1010005EA 116 EUR
ENZOBMLWN1010005 ENZOBMLWN1010025
PNU-74654 ≥98% (durch HPLC)
PNU-74654
Catenin blocker
Anti-tumor agent that disrupts the canonical Wnt/β-catenin pathway via blockage of the protein-protein interaction between β-catenin and TCF.
Formel: C₁₉H₁₆N₂O₃
Molecular Weight: 320,3419 g/mol
MDL: MFCD12912438
CAS-Nummer: 113906-27-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d6fc5751a6e8727311f847f619dc3316e960710c&vid=dbdfab9ce3899ab0723ea2528ffc0036351bcc43&prid=web&clid=SAPDAM