Tranylcypromine hemisulphate ≥99%
Lieferant: ENZO LIFE SCIENCES
Synonyme:
SKF trans-355, 2-PCPA, trans-2-Phenylcyclopropanamine hemisulphate
BMLEI2170001
BMLEI2170005
ENZOBMLEI2170001EA
165
EUR
ENZOBMLEI2170001
ENZOBMLEI2170005
Tranylcypromine hemisulphate ≥99%
Tranylcypromine hemisulphate
Demethylase inhibitor
Inhibits prostacyclin synthase. Monoamine oxidase inhibitor. Effective small molecule inhibitor of histone demethylation. Inhibits the demethylase BHC110/LSD1 (IC50<2μM).
Formel:
C₁₈H₂₄N₂O₄S Molecular Weight: 364,46 g/mol |
MDL:
MFCD00079222 CAS-Nummer: 13492-01-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...