N-Butyldeoxynojirimycin hydrochloride ≥98% (durch TLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme: Miglustat HCl, (2R,3R,4R,5S)-1-Butyl-2-(hydroxymethyl)piperidine-3,4,5-triol hydrochloride
BMLSL2300005 BMLSL2300025
ENZOBMLSL2300005EA 161 EUR
ENZOBMLSL2300005 ENZOBMLSL2300025
N-Butyldeoxynojirimycin hydrochloride ≥98% (durch TLC)
N-Butyldeoxynojirimycin hydrochloride
A novel inhibitor of glucosylceramide synthase as well as glucosidase I and II. Inhibits glucosylceramide synthase without ceramide accumulation and cell death. It is a potent inhibitor of human immunodeficiency virus (HIV) replication and syncytium formation in vitro. Restores chloride current in epithelial CF cells.
  • White to off-white solid
Soluble in water
Formel: C₁₀H₂₁NO₄·HCl
Molecular Weight: 255.7 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00269940
CAS-Nummer: 210110-90-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=99f53856b1f856dd4f261ce99cc402bb08af220e&vid=ccad14a0bf2bf2a334dbb9af4e0359fbc2a9ed1b&prid=web&clid=SAPDAM