CL-82198 (MMP-13 inhibitor) ≥98% (durch TLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme: N-[4-(4-morpholinyl)butyl]-2-benzofurancarboxamide, CL-82198, N-(4-Morpholinobutyl)benzofuran-2-carboxamide
BMLEI3020005 BMLEI3020025
ENZOBMLEI3020005EA 84.7 EUR
ENZOBMLEI3020005 ENZOBMLEI3020025
CL-82198 (MMP-13 inhibitor) ≥98% (durch TLC)
CL-82198 (MMP-13 inhibitor)
A novel and selective inhibitor of MMP-13 (89% at 33 µM). Acts via binding to the S1' pocket of MMP-13 without metal chelation.
  • MMP inhibitor
White solid.
Soluble in water (25mg/ml).
Formel: C₁₇H₂₂N₂O₃
Molecular Weight: 302,37 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12828877
CAS-Nummer: 307002-71-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b64309f5643347c61a5c042fd751405b920fdb74&vid=72e825ea04d3685a86e3cf07f7a108540eae3fd0&prid=web&clid=SAPDAM