BML-284 ≥98% (durch TLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme: N4-(Benzo[d][1,3]dioxol-5-ylmethyl)-6-(3-methoxyphenyl)pyrimidine-2,4-diamine
BMLWN1050005 BMLWN1050025
ENZOBMLWN1050005EA 161 EUR
ENZOBMLWN1050005 ENZOBMLWN1050025
BML-284 ≥98% (durch TLC)
BML-284
Wnt activator
Wnt agonist that activates Wnt signaling in cellular assays and a Xenopus model. Induces β-catenin and TCF-dependent transcriptional activity in a dose-dependent manner (EC50=0.7μM). Wnt activation occured in a GSK-3β independent manner.
Formel: C₁₉H₁₈N₄O₃
Molecular Weight: 350.4 g/mol
MDL: MFCD09908087
CAS-Nummer: 853220-52-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=892e6873cb36fb92b1d3b88a180cc9e36b0545e8&vid=d388829435f09622eda4699276cad9c1f3aa7104&prid=web&clid=SAPDAM