Prostaglandin F2α ≥99% (durch TLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme: PGF2alpha, Prostaglandin F2alpha
BMLPG0080002 BMLPG0080010
ENZOBMLPG0080002EA 92.4 EUR
ENZOBMLPG0080002 ENZOBMLPG0080010
Prostaglandin F2α ≥99% (durch TLC)
Prostaglandin F2α
A major uterine luteolytic prostaglandin. Endogenous agonist of the prostanoid FP receptor (KD=1.0nM). Inhibits differentiation of 3T3-L1 preadipocytes.
  • Prostanoid receptor ligand
Low melting white solid or colorless oil.
Soluble in 100% ethanol (50mg/ml).
Formel: C₂₀H₃₄O₅
Molecular Weight: 354,5 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00135231
CAS-Nummer: 551-11-1