ML-9 ≥98% (durch TLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme: 1-(5-Chloronaphthalene-1-sulphonyl)-1H-hexahydro-1,4-diazepine hydrochloride, 1-((5-Chloronaphthalen-1-yl)sulphonyl)-1,4-diazepane hydrochloride
BMLEI1530010 BMLEI1530050
ENZOBMLEI1530010EA 110 EUR
ENZOBMLEI1530010 ENZOBMLEI1530050
ML-9 ≥98% (durch TLC)
ML-9
Selective inhibitor of MLC kinase (Ki=3.8 µM). Inhibits insulin-stimulated 2-deoxyglucose transport and PP-1 activation in adipocytes, possibly via an effect on MAP kinase or its activation. Useful PKB inhibitor (IC50=10-50 µM). Also known to inhibit PKA (IC50=20 µM) and S6 kinase (IC50=50 µM). Does not inhibit PI 3-kinase.
  • Myelin basic protein inhibitor
White solid.
Soluble in DMSO (10mg/ml) and ethanol/water (1:1, 10mg/ml).
Formel: C₁₅H₁₇N₂O₂SCl·HCl
Molecular Weight: 361,3 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00065525
CAS-Nummer: 105637-50-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4b94dc77233943f715d519eea7ba7e04bbe342b3&vid=524f5aa87f5ef5d5a731985088ad402e98df634f&prid=web&clid=SAPDAM