Ko143 ≥96% (durch HPLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme: (3S,6S,12aS)-1,2,3,4,6,7,12,12a-Octahydro-9-methoxy-6-(2-methylpropyl)-1,4-dioxopyrazino-[1',2':1,6]pyrido[3,4-b]indole-3-propanoic acid 1,1-dimethylethyl ester
BMLEI3960005 BMLEI3960025
ENZOBMLEI3960005EA 208 EUR
ENZOBMLEI3960005 ENZOBMLEI3960025
Ko143 ≥96% (durch HPLC)
Ko143
Analog of the fungal toxin fumitremorgin C. Potent and selective inhibitor of the breast cancer resistance protein multidrug transporter (BCRP, EC90=26nM). Highly effective for increasing the intracellular drug accumulation and reversing BCRP-mediated multidrug resistance yet has little activity against P-glycoprotein, multidrug resistance-associated protein (MRP1) and other known drug transporters. In mouse models Ko143 was able to restore the intracellular accumulation and abolish the directionality in net flux of dasatinib. It is a highly useful tool for the identification of drug transport processes in complex experimental systems.
  • BCRP inhibitor
White solid.
Soluble in DMSO (>25mg/ml) or 100% ethanol (>25mg/ml).
Formel: C₂₆H₃₅N₃O₅
Molecular Weight: 469,6 g/mol
Lagertemperatur: Gefrierschrank
CAS-Nummer: 461054-93-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0b718f5b1073a8bfa8957b26182d16bce6a68ceb&vid=8bcd8762d67fba000dd8c987da82c6750de8dcd8&prid=web&clid=SAPDAM