13,14-Dihydro-prostaglandin E1 ≥98% (durch TLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme: 13,14-Dihydro-PGE1
BMLPG0480001 BMLPG0480010
ENZOBMLPG0480001EA 132 EUR
ENZOBMLPG0480001 ENZOBMLPG0480010
13,14-Dihydro-prostaglandin E1 ≥98% (durch TLC)
13,14-Dihydro-prostaglandin E1
Circulating metabolite of PGE1 in man. Vasodilator equipotent with PGE12. Inhibits human platelet aggregation. Antimitotic activity.
  • Prostanoid receptor ligand
Colorless to pale yellow oil
Formel: C₂₀H₃₆O₅
Molecular Weight: 356,5 g/mol
Lagertemperatur: Ultra-Tiefkühlschrank (–80°C)
CAS-Nummer: 19313-28-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1a2a39ec7c04a2099769297d2fc9ecb968f5fc3c&vid=0769180a8a581f30bc613957ae2060b15402dde1&prid=web&clid=SAPDAM