cis-4,7,10,13,16,19-Docosahexaensäure 98, Oil_Colourless, ultrapure

Lieferant: ENZO LIFE SCIENCES

Synonyme: DHA, Docosa-4Z,7Z,10Z,13Z,16Z,19Z-Hexaenoic Acid

BMLFA0020100 BMLFA0021000
ENZOBMLFA0020100EA 86.9 EUR
ENZOBMLFA0020100 ENZOBMLFA0021000
cis-4,7,10,13,16,19-Docosahexaensäure 98, Oil_Colourless, ultrapure
cis-4,7,10,13,16,19-Docosahexaenoic acid

Polyunsaturated fatty acid.


Soluble in ethanol, hexane and methylene chloride.

Formel: C₂₂H₃₂O₂
Molecular Weight: 328.50 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00065722
CAS-Nummer: 6217-54-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=405d38db02874f4083895c0237a0f9afd14a3b10&vid=44c7fe7b7b208769f34a5de99a18dcb81478fae4&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Purity ≥98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR