Anti-IgG Donkey Polyclonal Antibody (CF®583R Features)

Lieferant: Biotium
20892-50UL 20892-500UL
20892-50ULEA 103 EUR
20892-50UL 20892-500UL
Anti-IgG Donkey Polyclonal Antibody (CF®583R Features)
Antikörper
This is a highly cross-adsorbed donkey anti-goat IgG (H L) secondary antibody labeled with our bright and photostable CF® Dyes. To minimise cross-reactivity, the antibody has been adsorbed against chicken, guinea pig, Syrian hamster, horse, human, mouse, rabbit, and rat serum.

  • Highly cross-adsorbed for specific staining with minimal background
  • Available in biotin and with 20 bright and photostable CF® dyes
  • Suitable for western, immunofluorescence, and immunohistology in FFPE tissues

CF®583R dyes are two spectrally similar red fluorescent dyes with exceptional brightness. These dyes may be excited with a 561 nm to 568 nm laser line and have an emission maximum around 609 nm.

CF®583R is a rhodamine-based fluorescent dye that serves as an alternative to Texas Red®. CF®583R could be used as an excellent energy acceptor in Förster resonance energy transfer (FRET) when paired with R-PE. CF®583R produce antibody conjugates with brighter fluorescence compared to spectrally similar dye. In addition, the exceptional water solubility of the dyes facilitate bioconjugation and better preserves biological specificity of the conjugates. CF®583provided in amine-reactive forms and with many other bioconjugates. Additional reactive dyes or conjugates may be added on request.

Abs/Em maxima: 586/609 nm.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF®583R
Epitope:
Host: Donkey
Isotype: IgG (H L)
Reactivity: Goat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR