Nω-Nitro-L-Arginin-Methylester -hydrochlorid ≥98% (durch TLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme: N-ω-Nitro-L-Arginine Methyl Ester Hydrochloride, L-NAME Hydrochloride, L-NAME HCl, H-Arg(NO2)-OMe.HCl, (S)-Methyl 2-amino-5-(3-nitroguanidino)pentanoate hydrochloride
ALX105003G005 ALX105003G025
ENZOALX105003G005EA 104 EUR
ENZOALX105003G005 ENZOALX105003G025
Nω-Nitro-L-Arginin-Methylester -hydrochlorid ≥98% (durch TLC)
Nω-Nitro-L-Arginin-Methylester -hydrochlorid
Inhibitor of nitric oxide synthase (NOS). More soluble than NG-Nitro-L-arginine. Suppresses the release of nitric oxide (NO) from endothelial cells. Has a different effect on the superoxide generation by NOS than L-NMMA.
  • NOS inhibitor
White to off-white solid.
Soluble in water (50mg/ml) or methanol (25g/ml).
Formel: C₇H₁₅N₅O₄
Molecular Weight: 269,69 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00039052
CAS-Nummer: 51298-62-5