Indometacin
Lieferant: ENZO LIFE SCIENCES
ALX270086G005
ALX270086G025
ENZOALX270086G005EA
55
EUR
ENZOALX270086G005
ENZOALX270086G025
Indometacin
Indomethacin
Formel:
C₁₉H₁₆ClNO₄ Molecular Weight: 357.80 g/mol |
MDL:
MFCD00057095 CAS-Nummer: 53-86-1 EINECS: 200-186-5 UN: 2811 ADR: 6.1 I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...