NS-2028 ≥98% (durch TLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme: 4H-8-Bromo-1,2,4-oxadiazolo(3,4-d)benz(b)(1,4)oxazin-1-one
ALX270210M001 ALX270210M005
ENZOALX270210M001EA 57.2 EUR
ENZOALX270210M001 ENZOALX270210M005
NS-2028 ≥98% (durch TLC)
NS-2028
A potent and selective, cell-permeable inhibitor of nitric oxide (NO)-sensitive soluble guanylyl cyclase (sGC). Inhibition is irreversible. Inhibits also the basal, the YC-1 and the coenhanced sGC activity. Complete inhibition of NO-induced relaxation in vascular and nonvascular smooth muscle at 1μM.
  • Guanylyl cyclase inhibitor
White to off-white crystalline powder.
Soluble in DMSO or 100% ethanol; insoluble in water.
Formel: C₉H₅BrN₂O₃
Molecular Weight: 269,1 g/mol
Lagertemperatur: Gefrierschrank
CAS-Nummer: 204326-43-2