Tyrphostin AG 1024 ≥98% (durch HPLC und UV-Methode)
Lieferant: ENZO LIFE SCIENCES
Synonyme: 2-(3-Bromo-5-(tert-butyl)-4-hydroxybenzylidene)malononitrile, AG1024
ALX270217M001 ALX270217M005
ENZOALX270217M001EA 119 EUR
ENZOALX270217M001 ENZOALX270217M005
Tyrphostin AG 1024 ≥98% (durch HPLC und UV-Methode)
Tyrphostin AG 1024
Specific inhibitor of insulin-like growth factor-1 (IGF-1) and insulin receptor kinases. Induces apoptosis by downregulation of the expression of phospho-Akt1 and Bcl-2. Also increases Bax, p53 and p21 expression.
  • IGF-1 inhibitor
Yellow powder.
Soluble in acetonitrile (5mg/ml), DMSO (8mg/ml), dichloromethane (23mg/ml) or methanol (5mg/ml); insoluble in 100% ethanol.
Formel: C₁₄H₁₃BrN₂O
Molecular Weight: 305.2 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD02179365
CAS-Nummer: 65678-07-1