Daphnetin ≥97%
Lieferant: ENZO LIFE SCIENCES
Synonyme: 7,8-Dihydroxy-2H-chromen-2-one, 7,8-Dihydroxycoumarin
ALX270281M005 ALX270281M025
ENZOALX270281M005EA 49.5 EUR
ENZOALX270281M005 ENZOALX270281M025
Daphnetin ≥97%
Daphnetin
Protein kinase inhibitor
Inhibitor of protein kinases. EGF receptor tyrosine kinase (IC50=7.87 µM), PKA (IC50=9.33 µM), PKC (IC50=25.01 µM). Antioxidant. Antimalarial agent.
Formel: C₉H₆O₄
Molecular Weight: 178.14 g/mol
MDL: MFCD00016977
CAS-Nummer: 486-35-1