PGP-4008 ≥93%
Lieferant: ENZO LIFE SCIENCES
Synonyme:
N-(1-Benzyl-2,3-dihydro-1H-pyrrolo[2,3-b]quinolin-4-yl)-2-phenylacetamide
ALX270290M002
ALX270290M010
ENZOALX270290M002EA
198
EUR
ENZOALX270290M002
ENZOALX270290M010
PGP-4008 ≥93%
PGP-4008
Selective P-glycoprotein (Pgp) inhibitor. Effectively reverses Pgp mediated drug resistance in NCI/ADR cells, but has no effect on the drug sensitivities of MCF-7/VP cells that overexpress MRP1.
- P-glycoprotein inhibitor
White to off-white powder.
Soluble in methanol, 100% ethanol (3mg/ml) or DMSO (20mg/ml).
Formel:
C₂₆H₂₃N₃O Molecular Weight: 393.5 g/mol Lagertemperatur: Gefrierschrank |
CAS-Nummer:
365565-02-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...