Finasterid ≥98% (durch UV-Methode)
Lieferant: ENZO LIFE SCIENCES
Synonyme: N-tert-Butyl-3-oxo-4-aza-5alpha-androst-1-en-17beta-carboxamide
ALX270491M100 ALX270491M500
ENZOALX270491M100EA 129 EUR
ENZOALX270491M100 ENZOALX270491M500
Finasterid ≥98% (durch UV-Methode)
Finasteride
5alpha-reductase inhibitor
Inhibitor of 5α-reductase, the enzyme which converts testosterone to the more potent androgen, 5α-dihydrotestosterone. Used as a treatment in benign prostatic hyperplasia and prostate cancer. Also used to treat male pattern hair loss.
Formel: C₂₃H₃₆N₂O₂
Molecular Weight: 372,55 g/mol
MDL: MFCD00869737
CAS-Nummer: 98319-26-7