D-myo-Inositol-1,4,5-trisphosphate, hexapotassium salt ≥98% (durch TLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme: Ins(1,4,5)P3.6K, IP3
ALX307007M001 ALX307007M005
ENZOALX307007M001EA 159 EUR
ENZOALX307007M001 ENZOALX307007M005
D-myo-Inositol-1,4,5-trisphosphate, hexapotassium salt ≥98% (durch TLC)
D-myo-Inositol-1,4,5-trisphosphate, hexapotassium salt
Product of phosphatidylinositol phospholipid hydrolysis.
Causes release of calcium from intracellular stores.
Hygroscopic and soluble in water.
Formel: C₆H₁₂K₃O₁₅P₃
Molecular Weight: 648,6 g/mol
Lagertemperatur: Gefrierschrank
CAS-Nummer: 103476-24-0
Bestellen
Spezifikation Testergebnisse
Purity ≥98% (TLC)