Okadasäure Ammoniumsalz

Lieferant: ENZO LIFE SCIENCES

Synonyme: Halochondrine A ammonium salt, OA

ALX350010C100 ALX350010M001
ENZOALX350010C100EA 330 EUR
ENZOALX350010C100 ENZOALX350010M001
Okadasäure Ammoniumsalz
Okadaic acid ammonium salt
Formel: C₄₄H₆₇O₁₃·NH₄
Molecular Weight: 822.03 g/mol
MDL: MFCD00153856
CAS-Nummer: 155716-06-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR