Okadasäure Kaliumsalz
Lieferant: ENZO LIFE SCIENCES
Synonyme:
Halochondrine A potassium salt, OA potassium salt, 9,10-deepithio-9,10-didehydroacanthifolicin potassium salt
ALX350063C050
ALX350063C100
ALX350063M001
ENZOALX350063C050EA
165
EUR
ENZOALX350063C050
ENZOALX350063C100
ENZOALX350063M001
Okadasäure Kaliumsalz
Okadaic acid potassium salt
Formel:
C₄₄H₆₇O₁₃·K Molecular Weight: 843,11 g/mol |
MDL:
MFCD00214356 CAS-Nummer: 155751-72-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...