Betulinsäure ≥99%, hochrein

Lieferant: ENZO LIFE SCIENCES

Synonyme: (1R,3aS,5aR,5bR,7aR,9S,11aR,11bR,13aR,13bR)-9-Hydroxy-5a,5b,8,8,11a-pentamethyl-1-(prop-1-en-2-yl)icosahydro-1H-cyclopenta[a]chrysene-3a-carboxylic acid, 3beta-Hydroxy-20(29)-lupaene-28-oic Acid

ALX350277M005 ALX350277M025
ENZOALX350277M005EA 51.7 EUR
ENZOALX350277M005 ENZOALX350277M025
Betulinsäure ≥99%, hochrein
Betulinic acid

Antitumor and anti-HIV agent


Antitumor and anti-HIV agent. Induces apoptosis by activating mitochondrial permeability transition (MPT). Inhibits NF-κB activation and NF-κB-regulated gene expression induced by carcinogens and inflammatory stimuli. Decreases expression of Bcl-2 and cyclin D1. Potent proteasome activator. Triggers CD95- and p53-independent apoptosis via activation of caspases. It induces mitochondrial permeability pore opening and the release of cytochrome c and apoptosis-inducing factor. Displays anti-HIV activity. EC50 for inhibion of HIV-1 replicaton in H9 lymphocytes: 1.4µM.


Isolated from Platanus acerifolia (plane) tree bark.

Formel: C₃₀H₄₈O₃
Molecular Weight: 456.71 g/mol
MDL: MFCD00009619
CAS-Nummer: 472-15-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR