Betulinsäure ≥95%

Lieferant: ENZO LIFE SCIENCES

Synonyme: (1R,3aS,5aR,5bR,7aR,9S,11aR,11bR,13aR,13bR)-9-Hydroxy-5a,5b,8,8,11a-pentamethyl-1-(prop-1-en-2-yl)icosahydro-1H-cyclopenta[a]chrysene-3a-carboxylic acid, 3beta-Hydroxy-20(29)-lupaene-28-oic Acid

ALX350298G001 ALX350298M500
ENZOALX350298G001EA 574 EUR
ENZOALX350298G001 ENZOALX350298M500
Betulinsäure ≥95%
Betulinic acid

Proteasome activator


Antitumor and anti-HIV agent. Induces apoptosis by activating mitochondrial permeability transition (MPT). Inhibits NF-κB activation and NF-κB-regulated gene expression induced by carcinogens and inflammatory stimuli. Decreases expression of Bcl-2 and cyclin D1. Potent proteasome activator.


Isolated from Platanus acerifolia (plane tree) bark.

Formel: C₃₀H₄₈O₃
Molecular Weight: 456.71 g/mol
MDL: MFCD00009619
CAS-Nummer: 472-15-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR