MGD sodium salt monohydrate ≥98% (durch HPLC)
Lieferant: ENZO LIFE SCIENCES
Achtung
Synonyme: N-(Dithiocarbamoyl)-N-methyl-D-glucamine.Na.H₂O
ALX400014M050 ALX400014M250
ENZOALX400014M050EA 105 EUR
ENZOALX400014M050 ENZOALX400014M250
MGD sodium salt monohydrate ≥98% (durch HPLC)
MGD sodium salt monohydrate
Spin trapping reagent.
Together with FeSO₄ MGD is a useful component for the formation of the MGD2-Fe²⁺ complex, which is an excellent nitric oxide (NO) spin-trapping reagent. The MGD2-Fe²⁺ complex is quite unstable, especially in the presence of dissolved oxygen. Thus, the complex should be used immediately after being made. An excess (usually 5-fold excess), of MGD to Fe²⁺ is used for making the complex with FeSO₄ to give a more stable complex solution. Acidic conditions should be avoided because dithiocarbamate tends to decompose forming toxic carbon disulfide.
Soluble in water.
Formel: C₈H₁₆NO₅S₂Na·H₂O
Molecular Weight: 311.3 g/mol
Lagertemperatur: Kühlschrank
CAS-Nummer: 91840-27-6
Bestellen
Spezifikation Testergebnisse
Purity ≥98% (HPLC)