S-Nitrosoglutathione (GSNO) ≥98% (durch TLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme: (S)-2-Amino-5-(((R)-1-((carboxymethyl)amino)-3-(nitrosothio)-1-oxopropan-2-yl)amino)-5-oxopentanoic acid, GSNO, Nitrosoglutathione, S-Nitroso-L-Glutathione
ALX420002M025 ALX420002M100
ENZOALX420002M025EA 136 EUR
ENZOALX420002M025 ENZOALX420002M100
S-Nitrosoglutathione (GSNO) ≥98% (durch TLC)
S-Nitrosoglutathione (GSNO)
Nitric oxide donor
NO donor which spontaneously releases NO under physiological conditions. Half-life of 159 hours under physiological conditions. Selectively inhibits platelet activation and displays cardioprotective effects. Inhibits NF-kappaB activation. Inhibits endothelial cell proliferation and induces apoptosis in T cells.
Formel: C₁₀H₁₆N₄O₇S
Molecular Weight: 336.319 g/mol
MDL: MFCD00214341
CAS-Nummer: 57564-91-7