YC-1 ≥98% (durch HPLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme: (5-(1-Benzyl-1H-indazol-3-yl)furan-2-yl)methanol, Lificiguat
ALX420025M001 ALX420025M005
ENZOALX420025M001EA 49.5 EUR
ENZOALX420025M001 ENZOALX420025M005
YC-1 ≥98% (durch HPLC)
YC-1
Guanylyl cyclase activator
Nitric oxide (NO) independent, superoxide-sensitive stimulator of soluble guanylyl cyclase (sGC), EC50=18.6μM. Induces concentration-dependent increase in cGMP levels in cultured cells and in rabbit aortic rings. Inhibits platelet adhesion to collagen. Non-specific phosphodiesterase inhibitor. YC-1 inhibits HIF-alpha activity in tumors resulting in blocked angiogenesis and an inhibition of tumor growth. Inhibits Na+ channel.Binds to [3H]BW202W92.
Formel: C₁₉H₁₆N₂O₂
Molecular Weight: 304,349 g/mol
MDL: MFCD06407798
CAS-Nummer: 170632-47-0