Imiquimod

Lieferant: ENZO LIFE SCIENCES

Synonyme: 1-Isobutyl-1H-imidazo[4,5-c]quinolin-4-amine, 4-Amino-1-isobutyl-1H-imidazo[4,5-c]quinoline

ALX420039M100 ALX420039M250
ENZOALX420039M100EA 99 EUR
ENZOALX420039M100 ENZOALX420039M250
Imiquimod
Imiquimod

TLR7 ligand


Topical immune response modifier that inhibits angiogenesis. Up-regulates IL-18 and down-regulates MMP-9 through recognition of Toll-like receptor 7 (TLR7) and subsequent activation of MyD88-dependent pathway.

Formel: C₁₄H₁₆N₄
Molecular Weight: 240,31 g/mol
MDL: MFCD00866946
CAS-Nummer: 99011-02-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR